Learn More
Ace Glass, Inc. 50mm x 61cm chromatography column
SDP
Catalog No. 50972557
Encompass_Preferred
50mm X 61cm chromatography column with 2mm bore. Also features 1:5 taper solid PTFE stopcock plug and porosity B (70-100m) fritted disc sealed in.
Catalog No. 50-972-557 Supplier Ace Glass, Inc. Supplier No. 588940
50mm X 61cm chromatography column with 2mm bore. Also features 1:5 taper solid PTFE stopcock plug and porosity B (70-100m) fritted disc sealed in.