Learn More
Malvern Panalytical 50Nm Latex Std 3Ml
Supplier:  Malvern Panalytical 10047286
Encompass
50nm size calibrated Standards, highly uniform polystyrene spheres, packaged as aqueous suspensions.
Catalog No. NC1379640