Learn More
Medex Supply Abbott FreeStyle Lite Test Strips, 50/bx
SDP
Catalog No. NC1370524
Encompass
FreeStyle Lite test strips are specially designed for fast, easy testing and are intended to be used with FreeStyle Lite and FreeStyle Freedom Lite blood glucose monitoring systems only. The patented ZipWik tabs break the surface of the blood sample for instant wicking action. Plus, there’s no coding required.
This item is not returnable. View return policy
Catalog No. NC1370524 Supplier Medex Supply Supplier No. ADE70822
FreeStyle Lite test strips are specially designed for fast, easy testing and are intended to be used with FreeStyle Lite and FreeStyle Freedom Lite blood glucose monitoring systems only. The patented ZipWik tabs break the surface of the blood sample for instant wicking action. Plus, there’s no coding required.