Learn More
LabScientific 50-place premium microscope slide box. (Blue) Durable ABS plastic box closes securely with a rust-resistant nickel-plated clasp and pin hinges. Securely stores slides in grooved, labeled slots.
Catalog No. NC1460175
Encompass
50-place premium microscope slide box. (Blue) Durable ABS plastic box securely stores slides in grooved, labeled slots and closes tightly. The molded feet enable secure stacking for easy transport and storage. Both the lining and the slots are numbered to correspond to the slide inventory sheet. Dimensions: L x W x H: 209 x 86 x 35 mm
Catalog No. NC1460175 Supplier LabScientific Supplier No. SB50B
50-place premium microscope slide box. (Blue) Durable ABS plastic box securely stores slides in grooved, labeled slots and closes tightly. The molded feet enable secure stacking for easy transport and storage. Both the lining and the slots are numbered to correspond to the slide inventory sheet. Dimensions: L x W x H: 209 x 86 x 35 mm