Learn More
Aqua Solutions 50PPM TOC STANDARD 500ML
SDP
Catalog No. NC9566097
Encompass_Preferred
Total Organic Carbon Standard 50.0 Ppm Aqueous Solution, Prepared To Epa 415 Std Methods (500ml)
Catalog No. NC9566097 Supplier Aqua Solutions Supplier No. 9571500ML
Total Organic Carbon Standard 50.0 Ppm Aqueous Solution, Prepared To Epa 415 Std Methods (500ml)