Learn More
Athens Research And Technology Inc 50UG APO E HUMAN PLASMA
Supplier:  Athens Research And Technology Inc 161612050050UG
Encompass_Preferred
Apolipoprotein E, Human Plasma, VLDL: 50 UG; Frozen in 50 mM NH4HCO3, pH 8.0; MW 34,000; Purity: > =95% by SDS-PAGE. This native protein is prepared and purified from fresh, non-frozen human plasma in the Athens Research & Tech lab.
Catalog No. 50-489-654