Learn More
Alkali Scientific Alkali Scientific™ 50uL Beckman™ Biomek FX/NX, Multimek and i Series Compatible Robotic Automation Black Conductive Tips (P50), 96-Well Head/Tip Format, Low Retention, Sterile, Non-Filtered, 5 Packs of 960, 4800/Case
SDP
Catalog No. 503026946
Encompass_Preferred
Alkali Scientific™ offers a comprehensive line of tips for Beckman Coulter automation systems. These black conductive tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Tips are certified RNase-/DNase-free and nonpyrogenic. Designed for Beckman Coulter Biomek FX, Biomek NX, Biomek 3000 and Multimek AP-96 and i Series. Filtered tips reduce the chance of sample cross-contamination from aerosols. Like all our tips, the P50s are made of 100% polypropylene, are manufactured to ensure they are straight and leak-proof, and are guaranteed to be free of DNA, DNase/RNase, PCR inhibition, pyrogen/endotoxin and trace metals. DNase, RNase, and pyrogen-free. Crafted from high-grade polypropylene ISO 9001/13485, ISO 14001, and CE certified
Catalog No. 50-302-6946 Supplier Alkali Scientific Supplier No. FXBS50CS
Alkali Scientific™ offers a comprehensive line of tips for Beckman Coulter automation systems. These black conductive tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Tips are certified RNase-/DNase-free and nonpyrogenic. Designed for Beckman Coulter Biomek FX, Biomek NX, Biomek 3000 and Multimek AP-96 and i Series. Filtered tips reduce the chance of sample cross-contamination from aerosols. Like all our tips, the P50s are made of 100% polypropylene, are manufactured to ensure they are straight and leak-proof, and are guaranteed to be free of DNA, DNase/RNase, PCR inhibition, pyrogen/endotoxin and trace metals. DNase, RNase, and pyrogen-free. Crafted from high-grade polypropylene ISO 9001/13485, ISO 14001, and CE certified