Learn More
Trajan Scientific Hamilton 50l, X-Type Syringe FN GT
Supplier:  Trajan Scientific 100.SYRXG5022S3
Encompass_Preferred
50ul CTC X-Type Syringe, Fixed Needle, Gas Tight, PTFE, 22gs, pt3, special Needle coating CTC; Hamilton Part Number 204379
Catalog No. 50-635-418