Optimize your liquid handling processes with our 50 L CyBio-Style Clear Robotic Automation Tips designed for seamless compatibility with CyBio FeliX Well Flex and Vario Liquid Workstations Each box contains 768 tips (3840 tips per case) ensuring a steady supply for your high-throughput operations Key Features Precision Engineering Tailored for the Cybi-Well Family including Cybi-Well Cybi-Well Vario and Cybi-WellFlex these tips deliver consistent and accurate liquid handling for all automation applications Low Retention Design Our advanced low retention technology reduces sample loss ensuring optimal performance and reproducibility in every experiment Sterile and Clean Manufactured in a 100000-class cleanroom (Class 8) these tips are DNase RNase and pyrogen free Each batch undergoes E-beam sterilization and SGS verification assuring purity and safety Versatile Application Ideal for various liquid handling tasks these tips are compatible with a 96-well head/tip format offering fle