Learn More
Tecan 50 µL Disposable Tips - Non-filtered, Sterile, Single Stack, 40 Tip Boxes x 384-Tips
Supplier:  Tecan 30138088
Encompass_Preferred
MultiChannel Arm™ (MCA) 384 Disposable Tips are precision manufactured to offer high accuracy (the ability to dispense the desired volume) and low CVs (the variation of pipetting results), even at low volumes. Manufacturing is fully automated in a controlled environment, using state-of-the-art production processes, integrated quality control procedures and premium raw materials.
Catalog No. NC2944903