Learn More
Trajan Scientific 50UL EVOL SYRINGE
SDP
Catalog No. 502530071
Encompass_Preferred
SGE eVol Handheld Automated Analytical Syringe, eVol Analytical 25 ga, Volume: 50 uL, Needle Point Style: Bevel, Quantity: Each, Length Needle: 50 mm, 1.96 in., Needle Gauge: 25 G, Diameter Inner Needle: 0.007 in., 0.2 mm, Diameter Outer Needle: 0.019 in., 0.5 mm
Catalog No. 50-253-0071 Supplier Trajan Scientific Supplier No. 2910022
SGE eVol Handheld Automated Analytical Syringe, eVol Analytical 25 ga, Volume: 50 uL, Needle Point Style: Bevel, Quantity: Each, Length Needle: 50 mm, 1.96 in., Needle Gauge: 25 G, Diameter Inner Needle: 0.007 in., 0.2 mm, Diameter Outer Needle: 0.019 in., 0.5 mm