Learn More
Thomas Scientific Liquid Level Sensing Tips, Max Vol.: 50æl, Fits: Tecan, Qty: 50 racks of 96
SDP
Catalog No. 50819309
Encompass
Carbon black conductive polypropylene For Hamilton, Packard MultiProbe, Rosys, Qiagen, Tecan Genesis, Zymark and others Conductive material assists automated liquid handling systems in detecting the presence or absence of sufficient liquid. Packed in heavy duty racks, shrink wrapped and sterilized.
Catalog No. 50-819-309 Supplier Thomas Scientific Supplier No. 1158Q14
Carbon black conductive polypropylene For Hamilton, Packard MultiProbe, Rosys, Qiagen, Tecan Genesis, Zymark and others Conductive material assists automated liquid handling systems in detecting the presence or absence of sufficient liquid. Packed in heavy duty racks, shrink wrapped and sterilized.