Learn More
Alkali Scientific 50uL Hamilton STAR/STARlet Nim
SDP
Catalog No. 503027251
Encompass_Preferred
Alkali Scientificautomatic black conductive 50 L HamiltonSTAR/STARlet Nimbus and Microlab STARplus compatible robotic automation CO-RE style tips are crafted from high-quality polypropylene (PP) and treated with a specialized process to enhance hydrophobicity These black conductive tips are precisely engineered for specific applications providing enhanced efficiency and consistency in your work Material Manufactured from medical-grade high-tenacity polypropylene (PP) carefully selected and imported to meet strict industry standards ensuring consistent material quality between batches Production Environment Produced in 100000-class cleanroom conditions to ensure the absence of DNA RNA protease and endotoxins Each product is assigned a batch number for traceability improving experimental precision and minimizing product variability Surface Technology The unique processing ensures that the tips have a smooth inner surface greatly reducing liquid retention and improving transfer accu
Catalog No. 50-302-7251 Supplier Alkali Scientific Supplier No. HBFT1050
Alkali Scientificautomatic black conductive 50 L HamiltonSTAR/STARlet Nimbus and Microlab STARplus compatible robotic automation CO-RE style tips are crafted from high-quality polypropylene (PP) and treated with a specialized process to enhance hydrophobicity These black conductive tips are precisely engineered for specific applications providing enhanced efficiency and consistency in your work Material Manufactured from medical-grade high-tenacity polypropylene (PP) carefully selected and imported to meet strict industry standards ensuring consistent material quality between batches Production Environment Produced in 100000-class cleanroom conditions to ensure the absence of DNA RNA protease and endotoxins Each product is assigned a batch number for traceability improving experimental precision and minimizing product variability Surface Technology The unique processing ensures that the tips have a smooth inner surface greatly reducing liquid retention and improving transfer accu