Learn More
Alkali Scientific 50uL Hamilton STAR/STARlet Nim
SDP
Supplier:  Alkali Scientific HST1050
Encompass
Alkali Scientificautomatic transparent 50 L HamiltonSTAR/STARlet Nimbus and Microlab STARplus compatible robotic automation CO-RE style tips are crafted from high-quality polypropylene (PP) and treated with a special process to enhance hydrophobicity These transparent tips are meticulously designed for precise applications ensuring greater efficiency and repeatability in your work Material Made from medical-grade high-tenacity polypropylene (PP) sourced and selected according to strict industry standards to guarantee material consistency between batches Production Environment Manufactured in cleanroom conditions (100000-level purification) to prevent contamination from DNA RNA protease and endotoxins Every box is assigned a unique batch number for traceability improving experimental accuracy and minimizing variability Surface Technology Advanced processing ensures a smooth inner surface significantly reducing liquid retention and improving pipetting precision Structural Testing
Catalog No. 50-302-7412