Learn More
Trajan Scientific L-MARK Brand 50l Syringe, FN, GT, 22gs, NDL Length 51mm, Pt. Style 3, Plunger Tip PTFE, Single
Catalog No. 50635313
Encompass_Preferred
L-MARK Brand Syringe, 50-L, FN, GT, PTFE, 22s, Pt. 3; Syringe Adapter: PAL.SA050 Made in Germany.
Catalog No. 50-635-313 Supplier Trajan Scientific Supplier No. LMK.2620617
L-MARK Brand Syringe, 50-L, FN, GT, PTFE, 22s, Pt. 3; Syringe Adapter: PAL.SA050 Made in Germany.