Learn More
Ace Glass, Inc. 50 microliter gas-tight, fixed needle syringe
SDP
Catalog No. 50972627
Encompass
50 microliter gas-tight, fixed needle syringe. Syringe has a PTFE coated plunger with a precision PTFE tip for a leak-tight seal. Designed for highest performance in such applications as liquid or gas chromatography, handling of corrosive gases and liquids, radioactive materials and sterile solutions. Accuracy and reproducibility of plus or minus 1%.
Catalog No. 50-972-627 Supplier Ace Glass, Inc. Supplier No. 593102
50 microliter gas-tight, fixed needle syringe. Syringe has a PTFE coated plunger with a precision PTFE tip for a leak-tight seal. Designed for highest performance in such applications as liquid or gas chromatography, handling of corrosive gases and liquids, radioactive materials and sterile solutions. Accuracy and reproducibility of plus or minus 1%.