Learn More
Thermo Scientific Vanquish Pump Mixers | | 1 EA
Supplier:  Thermo Scientific 6044.5026
Encompass
Biocompatible; Designed for use with Thermo Scientific Vanquish systems; Ensures optimal mixing of HPLC eluent; Suitable for mixing difficult agents such as TFA; Provides high mixing efficiency for ultrahigh resolution and ultrafast separations; Achieves smooth baseline with high signal-to-noise ratios; Accurate peak identification; Reliable performance over a wide application range with varying solvent ratios, solvent miscibilities, flow rates
Catalog No. NC1811270