Learn More
Alkali Scientific 50uL PerkinElmer Revvity Janus
SDP
Catalog No. 503028340
Encompass_Preferred
Alkali Scientificdesigns its PerkinElmer-compatible automation tips to ensure flawless compatibility with PerkinElmer Janus Varispan liquid handlers Our tips require no separate labware definitions or modifications to PerkinElmer Janus automation control software This conductive black tip in a 96-format is fully interchangeable with the original PerkinElmer P200 automation pipette tip and is also compatible with Sciclone Zephyr Evolution P3 RapidPlate MiniTrak and MultiProbe II HT/ET systems Designed for precise volume transfers up to 200 L the PerkinElmer P200 conductive black tip delivers exceptional accuracy and flexibility Its capacity and length accommodate diverse protocol needs These tips are suitable for use with both 96-format and 384-format plates and are compatible with Janus Varispan pipetting arms Alkali Scientificproduces its PerkinElmer Janus-compatible P200 tips using high-quality virgin polypropylene Advanced polishing techniques minimize liquid retention witho
Catalog No. 50-302-8340 Supplier Alkali Scientific Supplier No. PEFT1050
Alkali Scientificdesigns its PerkinElmer-compatible automation tips to ensure flawless compatibility with PerkinElmer Janus Varispan liquid handlers Our tips require no separate labware definitions or modifications to PerkinElmer Janus automation control software This conductive black tip in a 96-format is fully interchangeable with the original PerkinElmer P200 automation pipette tip and is also compatible with Sciclone Zephyr Evolution P3 RapidPlate MiniTrak and MultiProbe II HT/ET systems Designed for precise volume transfers up to 200 L the PerkinElmer P200 conductive black tip delivers exceptional accuracy and flexibility Its capacity and length accommodate diverse protocol needs These tips are suitable for use with both 96-format and 384-format plates and are compatible with Janus Varispan pipetting arms Alkali Scientificproduces its PerkinElmer Janus-compatible P200 tips using high-quality virgin polypropylene Advanced polishing techniques minimize liquid retention witho