Learn More
Alkali Scientific 50uL PerkinElmer Revvity Janus
SDP
Catalog No. 503028957
Encompass_Preferred
Alkali Scientificdesigns its PerkinElmer automation tips for seamless compatibility with PerkinElmer Janus Varispan liquid handlers Using our PerkinElmer Janus-compatible automation tips eliminates the need for a separate labware definition and there is no need to modify the PerkinElmer Janus automation control software protocols This 96-format conductive black tip is fully interchangeable with the original PerkinElmer P50 automation pipette tip These tips are also compatible with Sciclone Zephyr Evolution P3 RapidPlate MiniTrak and MultiProbe II HT/ET systems ApplicationsThe PerkinElmer P50 conductive black tip is designed for volume transfers up to 50 L offering excellent precision and accuracy Its volume capacity and length provide flexibility for designing a wide range of protocols These tips are compatible with both 96-format and 384-format plates and fit Janus Varispan pipetting arms Alkali Scientificmanufactures its PerkinElmer Janus-compatible P50 tips from high-qualit
Catalog No. 50-302-8957 Supplier Alkali Scientific Supplier No. PEBST1050
Alkali Scientificdesigns its PerkinElmer automation tips for seamless compatibility with PerkinElmer Janus Varispan liquid handlers Using our PerkinElmer Janus-compatible automation tips eliminates the need for a separate labware definition and there is no need to modify the PerkinElmer Janus automation control software protocols This 96-format conductive black tip is fully interchangeable with the original PerkinElmer P50 automation pipette tip These tips are also compatible with Sciclone Zephyr Evolution P3 RapidPlate MiniTrak and MultiProbe II HT/ET systems ApplicationsThe PerkinElmer P50 conductive black tip is designed for volume transfers up to 50 L offering excellent precision and accuracy Its volume capacity and length provide flexibility for designing a wide range of protocols These tips are compatible with both 96-format and 384-format plates and fit Janus Varispan pipetting arms Alkali Scientificmanufactures its PerkinElmer Janus-compatible P50 tips from high-qualit