Learn More
Azzota Corp 10mm Pathlength (0.05Ml) Sub-Micro Cuvette Pathlength - 10mm Inside Width - 2X2.5mm Volume - 0.05Ml Dimensions - 45X12.5X12.5mm Have Black Walls Z-Height - 15mm Quartz (190Nm-2500Nm)
Catalog No. 507532897
Encompass_Preferred
10mm Pathlength (0.05ml) Sub-micro Cuvette Pathlength - 10mm Inside width - 2x2.5mm Volume - 0.05ml Dimensions - 45x12.5x12.5mm Have black walls Z-Height - 15mm (Option: Z=8.5mm is available) Quartz (190nm-2500nm)
Catalog No. 50-753-2897 Supplier Azzota Corp Supplier No. Q675
10mm Pathlength (0.05ml) Sub-micro Cuvette Pathlength - 10mm Inside width - 2x2.5mm Volume - 0.05ml Dimensions - 45x12.5x12.5mm Have black walls Z-Height - 15mm (Option: Z=8.5mm is available) Quartz (190nm-2500nm)