Learn More
Alkali Scientific 50uL Tecan MCA Robotic Automat
SDP
Supplier:  Alkali Scientific MCAST1050
Encompass_Preferred
Alkali Scientifichas designed its Tecan P50 MCA (MultiChannel Arm) tips for seamless compatibility with Fluent robotic liquid handlers When using our Fluent MCA-compatible automation tips no separate labware definition is required and Tecan automation control software protocols remain unchanged This 96-format tip is fully interchangeable with the original Tecan P50 automation pipette tip This 96-format tip is ideal for volume transfers up to 50 L (with filter) offering excellent flexibility in protocol design These Tecan P50 tips can be used to access both 96-format and 384-format plates and are compatible with all 96 MCA heads on Freedom EVO and Fluent liquid handlers Alkali Scientificmanufactures its Tecan Freedom EVO and Fluent-compatible P50 tips from high-quality virgin polypropylene Advanced polishing techniques are employed to minimize liquid retention with no use of releasing agents or additives Specifications Working volume 1 00 80 00 L 1 00 50 00 L (with filter)
Catalog No. 50-302-9440