Learn More
Fluotics Beckman Coulter Biomek iSeries FX NX compatible P50, non-sterile
Catalog No. NC2097984
Encompass_Preferred
Beckman Coulter Biomek iSeries FX NX compatible P50, non-sterile, working volume: 50 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: B85753
This item is not returnable. View return policy
Catalog No. NC2097984 Supplier Fluotics Supplier No. BFXI50.NS
Beckman Coulter Biomek iSeries FX NX compatible P50, non-sterile, working volume: 50 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: B85753