Learn More
Alkali Scientific 50uL Zymark-Style Clear Roboti
SDP
Catalog No. 503028178
Encompass_Preferred
Our robotic pipet tips are precisely engineered to match the specifications of original equipment manufacturer (OEM) tips ensuring perfect compatibility and a consistent fit with the leading liquid handling systems Our robotic pipet tips are designed for use with Zymark RapidPlate Zymark SciClone Tecan Te-MO Tecan Genmate and Tecan Aquariusautomated liquid handling systems Engineered to the precise dimensions of OEM tips these tips guarantee seamless automation compatibility and form an airtight seal with the robotic system Made from a specialized polymer they resist DNA RNA and protein adhesion They are manufactured using diamond-polished molds to eliminate any cavities or occlusions that may cause sample retention Additionally the tips are free from secondary substances like silicone which could potentially contaminate your samples Volume 50 L Color Clear Sterile Standard Style Certified RNase DNase and Pyrogen-Free Quantity 96 Tips/Rack 10 Racks/Unit 5 Units/Case
Catalog No. 50-302-8178 Supplier Alkali Scientific Supplier No. ZST1050
Our robotic pipet tips are precisely engineered to match the specifications of original equipment manufacturer (OEM) tips ensuring perfect compatibility and a consistent fit with the leading liquid handling systems Our robotic pipet tips are designed for use with Zymark RapidPlate Zymark SciClone Tecan Te-MO Tecan Genmate and Tecan Aquariusautomated liquid handling systems Engineered to the precise dimensions of OEM tips these tips guarantee seamless automation compatibility and form an airtight seal with the robotic system Made from a specialized polymer they resist DNA RNA and protein adhesion They are manufactured using diamond-polished molds to eliminate any cavities or occlusions that may cause sample retention Additionally the tips are free from secondary substances like silicone which could potentially contaminate your samples Volume 50 L Color Clear Sterile Standard Style Certified RNase DNase and Pyrogen-Free Quantity 96 Tips/Rack 10 Racks/Unit 5 Units/Case