Learn More
TRIBIOSCIENCE INC 50x Denhardt’s Solution 50ML
SDP
Catalog No. NC3344582
Encompass
50X Denhardt’s solution is a blocking reagent mostly applied for western, northern, and southern blot procedures. It prevents the probes non-specific binding to the membranes. The solution contains 1% Ficoll (type 400), 1% Polyvinylpyrrolidone and 1% bovine serum albumin (BSA).
Catalog No. NC3344582 Supplier TRIBIOSCIENCE INC Supplier No. TBS505350ML
50X Denhardt’s solution is a blocking reagent mostly applied for western, northern, and southern blot procedures. It prevents the probes non-specific binding to the membranes. The solution contains 1% Ficoll (type 400), 1% Polyvinylpyrrolidone and 1% bovine serum albumin (BSA).