Learn More
As One International Inc 50x Glass Blocking agent, 3 x 0.2 ml
Catalog No. 501656010
Encompass_Preferred
One extra challenge using the LightCycler instrument is that the PCR reagents can form precipitates on the glass capillary surface as the Real time PCR progresses. To prevent this event AS ONE offers a special reagent for blocking the glass capillaries during the Quantitative PCR reaction. The Glass blocking agent is a 50x solution (1 ul per 50 ul PCR reaction).
Catalog No. 50-165-6010 Supplier As One International Inc Supplier No. AO351413
One extra challenge using the LightCycler instrument is that the PCR reagents can form precipitates on the glass capillary surface as the Real time PCR progresses. To prevent this event AS ONE offers a special reagent for blocking the glass capillaries during the Quantitative PCR reaction. The Glass blocking agent is a 50x solution (1 ul per 50 ul PCR reaction).