Learn More
ILEX LIFE SCIENCES LLC 50X HAT SUPPLEMENT 100 ML
SDP
Catalog No. 502733720
Encompass_Preferred
PAN-Biotech HAT Supplement (50x), 100 ml, Cell Culture Media Supplement, Hypoxanthine + Aminopterin + Thymidine. DESCRIPTION: HAT Supplement (50x) is a liquid mixture of hypoxanthine, aminopterin, and thymidine used as a media supplement in cell culture applications. CONCENTRATION: 50x concentrated. STORAGE: -20°C. STERILE: Yes. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. COMPOSITION: Hypoxanthine: 680.50 mg/L; Thymidine: 388.00 mg/L; Aminopterin: 8.80 mg/L.
Catalog No. 50-273-3720 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0702100
PAN-Biotech HAT Supplement (50x), 100 ml, Cell Culture Media Supplement, Hypoxanthine + Aminopterin + Thymidine. DESCRIPTION: HAT Supplement (50x) is a liquid mixture of hypoxanthine, aminopterin, and thymidine used as a media supplement in cell culture applications. CONCENTRATION: 50x concentrated. STORAGE: -20°C. STERILE: Yes. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. COMPOSITION: Hypoxanthine: 680.50 mg/L; Thymidine: 388.00 mg/L; Aminopterin: 8.80 mg/L.