TAE (Tris-Acetate EDTA Buffer) buffer provides faster electrophoretic migration of linear DNA and better resolution of supercoiled DNA. TAE Buffer may be more suitable than TBE, for electrophoresis if DNA is to be recovered from the agarose and purified using commercially available DNA purification kits.
Applications: TAE buffer has a relatively low buffering capacity, therefore, periodic replacement of the buffer during prolonged electrophoresis is recommended.
Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase RNase and Protease Tested
Concentration: 2M Tris base and 0.05M disodium EDTA dihydrate
Max Shelf Life: 36 months from date of manufacture
Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.