Learn More
Quality Biological Inc TAE Buffer 50X, 4 Liters
SDP
Catalog No. 50146818
Encompass_Preferred
TAE buffer provides faster electrophoretic migration of linear DNA and better resolution of supercoiled DNA. ¿ TAE Buffer may be more suitable than TBE, for electrophoresis if DNA is to be recovered from the agarose and purified using commercially available DNA purification kits. ¿ Applications: TAE buffer has a relatively low buffering capacity, therefore, periodic replacement of the buffer during prolonged electrophoresis is recommended. ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ DNase RNase and Protease Tested ¿ Concentration: 2 M Tris base and 0.05 M disodium EDTA dihydrate ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months
Catalog No. 50-146-818 Supplier Quality Biological Inc Supplier No. 351008491
Add to Cart
Add to Cart
TAE buffer provides faster electrophoretic migration of linear DNA and better resolution of supercoiled DNA. ¿ TAE Buffer may be more suitable than TBE, for electrophoresis if DNA is to be recovered from the agarose and purified using commercially available DNA purification kits. ¿ Applications: TAE buffer has a relatively low buffering capacity, therefore, periodic replacement of the buffer during prolonged electrophoresis is recommended. ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ DNase RNase and Protease Tested ¿ Concentration: 2 M Tris base and 0.05 M disodium EDTA dihydrate ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months