Learn More
Teknova 50X TAE Solution. 1000mL, Non-Sterile.
SDP
Catalog No. 502388093
Encompass
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.
Catalog No. 50-238-8093 Supplier Teknova Supplier No. T5279
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.