Learn More
Rockland Immunochemicals 50X Tris-Acetate-EDTA (TAE) (2.0 M Tris-Acetate 0.05 M EDTA pH 8.3) - MB-020
SDP
Catalog No. NC9470178
Encompass_Preferred
Tris-Acetate-EDTA (TAE) is a buffer solution containing a mixture of Tris base, acetic acid and EDTA. TAE suitable for multiplemolecular biology applications including agarose electrophoresis for the separation of nucleic acids such as DNA and RNA.
Catalog No. NC9470178 Supplier Rockland Immunochemicals Supplier No. MB020
Add to Cart
Add to Cart
Tris-Acetate-EDTA (TAE) is a buffer solution containing a mixture of Tris base, acetic acid and EDTA. TAE suitable for multiplemolecular biology applications including agarose electrophoresis for the separation of nucleic acids such as DNA and RNA.