Learn More
Teknova 50X VB Salts Solution. 500mL, Sterile 8 pack.
SDP
Catalog No. 502387261
Encompass
1.0% Magnesium Sulfate, 10.0% Citric Acid,50.0% Dipotassium Phosphate, 17.5% Sodium Ammonium Phosphate
Catalog No. 50-238-7261 Supplier Teknova Supplier No. V25108PK
1.0% Magnesium Sulfate, 10.0% Citric Acid,50.0% Dipotassium Phosphate, 17.5% Sodium Ammonium Phosphate