Learn More
MOLTOX Molecular Toxicology Inc 50X VBE, Non-sterile, 500 mL/bottle, Store at Room Temperature
SDP
Catalog No. 50948755
Encompass
Vogel-Bonner medium E (VBE) and Vogel""s salts are used in the manufacture of Minimal Glucose Agar for use in the Ames assay. All MOLTOX reagents are accompanied by a GLP-compliant Formulation and Quality Control statement.
Catalog No. 50-948-755 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26550.047
Vogel-Bonner medium E (VBE) and Vogel""s salts are used in the manufacture of Minimal Glucose Agar for use in the Ames assay. All MOLTOX reagents are accompanied by a GLP-compliant Formulation and Quality Control statement.