Learn More
Ace Glass, Inc. 50x600mm epoxy coated chromatography column
SDP
Catalog No. 50978422
Encompass_Preferred
50x600mm high performance, non-packed, epoxy coated chromatography column. Simple to assemble and is safely operated to 150psi with safety shield.
Catalog No. 50-978-422 Supplier Ace Glass, Inc. Supplier No. 579554
50x600mm high performance, non-packed, epoxy coated chromatography column. Simple to assemble and is safely operated to 150psi with safety shield.