Learn More
Ted Pella Inc Petri Dish, 50 x 9mm, 20/pk
SDP
Catalog No. NC1246514
Encompass
Economical and optically clear polystyrene petri dish provides quality sample viewing without distortion. Tight lids prevent excess evaporation. For classroom studies, culturing of mycobacteria, aerosol testing, membrane filter and immunodiffusion techniques. Not for tissue culture application. A convenient 3-tab design on the lid provides secure handling for tissue culture use. Packaged in sleeves of 20, sterile.
Catalog No. NC1246514 Supplier Ted Pella Inc Supplier No. 14014
Economical and optically clear polystyrene petri dish provides quality sample viewing without distortion. Tight lids prevent excess evaporation. For classroom studies, culturing of mycobacteria, aerosol testing, membrane filter and immunodiffusion techniques. Not for tissue culture application. A convenient 3-tab design on the lid provides secure handling for tissue culture use. Packaged in sleeves of 20, sterile.