Learn More
Boca Scientific Inc GeneFix 2ml DNA/RNA Saliva Collectors, Pack of 50
Catalog No. NC1822414
Encompass
GeneFix is an easy-to-use saliva collection device that stabilizes DNA and viral RNA in tubes pre-filled with a non-toxic stabilization buffer. GFX-02 is developed for 2ml of saliva. The collector has a simple screw-on funnel for saliva delivery directly into the buffer. The saliva collection tube clearly indicates the volume of saliva required. Once in the buffer, DNA is stabilized for 5 years and viral RNA is stabilized for 5 days at room temperature, with high yields maintained. With leakproof collection tubes, GeneFix saliva collectors are fully specified for sample transport and storage.
Catalog No. NC1822414 Supplier Boca Scientific Inc Supplier No. GFX02/50
GeneFix is an easy-to-use saliva collection device that stabilizes DNA and viral RNA in tubes pre-filled with a non-toxic stabilization buffer. GFX-02 is developed for 2ml of saliva. The collector has a simple screw-on funnel for saliva delivery directly into the buffer. The saliva collection tube clearly indicates the volume of saliva required. Once in the buffer, DNA is stabilized for 5 years and viral RNA is stabilized for 5 days at room temperature, with high yields maintained. With leakproof collection tubes, GeneFix saliva collectors are fully specified for sample transport and storage.