Learn More
Chemglass Life Sciences 51-400 G.P.I. Glass Thread, 46mm OD Tubing, 125mm OAL
SDP
Catalog No. CG19015
Encompass_Preferred
51-400 G.P.I. Glass Thread, 46mm OD Tubing, 125mm OAL. Thread for replacement on apparatus using G.P.I. (Glass Packaging Institute) thread. PLEASE NOTE: GPI THREADS ARE SOLD IN PACKAGES ONLY.
Catalog No. CG19015 Supplier Chemglass Life Sciences Supplier No. CG19015
51-400 G.P.I. Glass Thread, 46mm OD Tubing, 125mm OAL. Thread for replacement on apparatus using G.P.I. (Glass Packaging Institute) thread. PLEASE NOTE: GPI THREADS ARE SOLD IN PACKAGES ONLY.