Learn More
Sino Biological Recombinant Mouse Pleiotrophin/PTN Protein
Supplier:  Sino Biological 51000MNAB100
Encompass_Preferred
A DNA sequence encoding the mouse PTN (P63089) (Met1-Asp168) was expressed.
Catalog No. 50-162-2743