Learn More
Sino Biological Recombinant Mouse TFPI2 Protein (His Tag)
Supplier:  Sino Biological 51002M08H100
Encompass_Preferred
A DNA sequence encoding the mouse TFPI2 (O35536) (Met 1-Lys 211) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2749