Learn More
Sino Biological Recombinant Mouse IL-12 p40/IL12B Protein (hFc Tag)
Supplier:  Sino Biological 51004M02H50
Encompass_Preferred
A DNA sequence encoding the mouse IL12B (P43432) (Met 1-Ser 335) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2754