Learn More
Sino Biological Recombinant Mouse IL-12 p40/IL12B Protein (His Tag)
Supplier:  Sino Biological 51004M08H50
Encompass_Preferred
A DNA sequence encoding the mouse IL12B (NP_001290173.1)(Met1-Ser335) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2756