Learn More
Sino Biological Recombinant Mouse Contactin 3 Protein (His Tag)
Supplier:  Sino Biological 51007M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CNTN3 (NP_032805.2) (Met1-Gly 1001) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2762