Learn More
Sino Biological Recombinant Mouse Prostatic Acid Phosphatase/ACPP Protein (His Tag)
Supplier:  Sino Biological 51018M08H100
Encompass_Preferred
A DNA sequence encoding the mouse ACPP isoform 1 (Q8CE08-1) (Met1-Arg 381) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2766