Learn More
Sino Biological Recombinant Mouse PDK4 Protein (His & GST Tag)
Supplier:  Sino Biological 51021M20B50
Encompass_Preferred
A DNA sequence encoding the mouse PDK4 (O70571) (Met1-Val412) was expressed with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-162-2771