Learn More
Sino Biological Recombinant Mouse AKT3 Protein (aa 106-479)
Supplier:  Sino Biological 51027MNCB50
Encompass_Preferred
A DNA sequence encoding the Mouse AKT3 (Q9WUA6-1) (Ala106-Glu479) was expressed and purified.
Catalog No. 50-162-2775