Learn More
Sino Biological Recombinant Mouse LAIR1 Protein (His Tag)
Supplier:  Sino Biological 51030M08H100
Encompass_Preferred
A DNA sequence encoding the mouse LAIR1 (Q8BG84-1) extracellular domain (Gln 22-Ser 133) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2778