Learn More
Sino Biological Recombinant Mouse GM-CSF/CSF2 Protein (hFc Tag)
Supplier:  Sino Biological 51048M01H50
Encompass_Preferred
A DNA sequence encoding the mouse CSF2 (P01587) (Ala18-Lys141) was was expressed with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-2795