Learn More
Sino Biological Recombinant Mouse CREG1 Protein (His Tag)
Supplier:  Sino Biological 51049M08H50
Encompass_Preferred
A DNA sequence encoding the mouse CREG (O88668) (Met1-Gln220) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2803