Learn More
Sino Biological Recombinant Mouse IL-2 Protein
Supplier:  Sino Biological 51061MNAE100
Encompass_Preferred
A DNA sequence encoding the soluble form of mouse IL2 (P04351) (Ala 21-Gln 169) was expressed, with an initial Met at the C-terminus.
Catalog No. 50-162-2806