Learn More
Sino Biological Recombinant Mouse ERBB4/Her4 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 51064M08H100
Encompass_Preferred
A DNA sequence encoding the mouse ERBB4 (Q99P91) (Met1-Leu652) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2809