Learn More
Sino Biological Recombinant Mouse IL-3 Protein, HPLC-verified
Supplier:  Sino Biological 51066MNAH50
Encompass_Preferred
A DNA sequence encoding the mouse IL3 (NP_034686.2) (Met1-Cys166) was expressed.
Catalog No. 50-162-2816